SY3H3D67BC1E Synthonix Corporation

1,2-Dibromo-4-methyl-5-nitrobenzene

  • MDL number MFCD23135744
  • Cas Number : 101580-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Light-yellow to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/44935.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/44935.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart