SY3H3D67C012 Synthonix Corporation

4,4-Dimethylpyrrolidine-3-carboxylic acid hydrochloride

  • MDL number MFCD19686958
  • Cas Number : 1351343-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46493.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46493.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart