SY3H3D67C013 Synthonix Corporation

2-(3,4-Difluorophenyl)morpholine hydrochloride

  • MDL number MFCD09832019
  • Cas Number : 1251033-05-2
  • Molecular Weight : 235.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to off-white powder or crystalsr
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54117.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54117.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart