SY3H3D67C016 Synthonix Corporation

2-(3,5-Dichloro-4-oxopyridin-1(4H)-yl)acetic acid

  • Cas Number : 56187-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/45009.html

Safety Information

MSDS https://synthonix.com/45009.html
Availability details for
Quantity
  • Availability
  • Add to Cart