SY3H3D67C019 Synthonix Corporation

6-(Difluoromethoxy)nicotinic acid

  • Cas Number : 1211535-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45012.html

Safety Information

MSDS https://synthonix.com/45012.html
Availability details for
Quantity
  • Availability
  • Add to Cart