SY3H3D67C028 Synthonix Corporation

2,6-Dibromotoluene

  • MDL number MFCD00013524
  • Cas Number : 69321-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Colorless to pale-yellow to brown liquid or solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45153.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45153.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart