SY3H3D67C033 Synthonix Corporation

3,5-Dichloro-6-ethylpyrazine-2-carboxylic acid

  • MDL number MFCD28506305
  • Cas Number : 1883347-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45239.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45239.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart