SY3H3D67C034 Synthonix Corporation

N,2-Dihydroxy-5-nitrobenzamide

  • MDL number MFCD00818219
  • Cas Number : 61494-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45258.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45258.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart