SY3H3D67C054 Synthonix Corporation

(Difluoromethoxy)benzene

  • MDL number MFCD00068184
  • Cas Number : 458-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45605.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45605.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart