SY3H3D67C05E Synthonix Corporation

3,6-Dimethylpyrazine-2-carbonitrile

  • MDL number MFCD21648456
  • Cas Number : 2435-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45730.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45730.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart