SY3H3D67C067 Synthonix Corporation

2,4-Dichloro-5-methylaniline

  • MDL number MFCD09833435
  • Cas Number : 17601-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Off-white to Gray or Yellow Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45787.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45787.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart