SY3H3D67C06A Synthonix Corporation

3,5-Dichloro-2-methylaniline

  • Cas Number : 62077-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45790.html

Safety Information

MSDS https://synthonix.com/45790.html
Availability details for
Quantity
  • Availability
  • Add to Cart