SY3H3D67C074 Synthonix Corporation

2,5-Difluoro-4-methylaniline

  • Cas Number : 878285-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/45809.html

Safety Information

MSDS https://synthonix.com/45809.html
Availability details for
Quantity
  • Availability
  • Add to Cart