SY3H3D67C075 Synthonix Corporation

4,5-Difluoro-2-methylaniline

  • MDL number MFCD08062378
  • Cas Number : 875664-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/45810.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/45810.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart