SY3H3D67C0A1 Synthonix Corporation

3,5-Diamino-2-hydroxybenzoic acid

  • MDL number MFCD09753462
  • Cas Number : 112725-89-0
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form White to light-yellow to gray to brown to black powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46030.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46030.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart