SY3H3D67C108 Synthonix Corporation

2,6-Dichloro-5-(trifluoromethyl)nicotinonitrile

  • Cas Number : 1319804-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46453.html

Safety Information

MSDS https://synthonix.com/46453.html
Availability details for
Quantity
  • Availability
  • Add to Cart