SY3H3D67C118 Synthonix Corporation

N,N-Diethyl-N-methylethanaminium tetrafluoroborate

  • MDL number MFCD08458747
  • Cas Number : 69444-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Lump, Waxy / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46614.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/46614.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart