SY3H3D67C119 Synthonix Corporation

1,2-Dimethyl-3-propyl-1H-imidazol-3-ium iodide

  • MDL number MFCD15072160
  • Cas Number : 218151-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46622.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46622.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart