SY3H3D67C11E Synthonix Corporation

3,6-Dichloro-4,5-diethylpyridazine

  • Cas Number : 107228-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46646.html

Safety Information

MSDS https://synthonix.com/46646.html
Availability details for
Quantity
  • Availability
  • Add to Cart