SY3H3D67C123 Synthonix Corporation

3,4-Dimethyl-5-nitroaniline

  • MDL number MFCD09880021
  • Cas Number : 64823-22-9
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/46671.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/46671.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart