SY3H3D67C128 Synthonix Corporation

(5,7-Dimethyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-yl)methanol

  • Cas Number : 54535-00-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46717.html

Safety Information

MSDS https://synthonix.com/46717.html
Availability details for
Quantity
  • Availability
  • Add to Cart