SY3H3D67C12C Synthonix Corporation

2,4-Dimethoxyphenol

  • Cas Number : 13330-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/46840.html

Safety Information

MSDS https://synthonix.com/46840.html
Availability details for
Quantity
  • Availability
  • Add to Cart