SY3H3D67C13D Synthonix Corporation

N1,3-Dimethylbenzene-1,2-diamine

  • Cas Number : 73902-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/46971.html

Safety Information

MSDS https://synthonix.com/46971.html
Availability details for
Quantity
  • Availability
  • Add to Cart