SY3H3D67C145 Synthonix Corporation

2-(2,4-Diaminophenoxy)ethanol sulfate

  • MDL number MFCD00035495
  • Cas Number : 70643-20-8
  • Molecular Weight : 266.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sulfuric acid compound with 2-(2,4-diaminophenoxy)ethanol (1:1)
Inchi Code 1S/C8H12N2O2.H2O4S/c9-6-1-2-8(7(10)5-6)12-4-3-11;1-5(2,3)4/h1-2,5,11H,3-4,9-10H2;(H2,1,2,3,4)
Inchi Key UUHBEKCQQLDQNG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Gray to Light purple to purple powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47003.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47003.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart