SY3H3D67C19A Synthonix Corporation

Dimethyl acetylsuccinate

  • MDL number MFCD00008448
  • Cas Number : 10420-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: Colorless - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47042.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47042.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart