SY3H3D67C19C Synthonix Corporation

5,8-Dioxa-10-azadispiro[2.0.4.3]undecane

  • Cas Number : 129321-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47059.html

Safety Information

MSDS https://synthonix.com/47059.html
Availability details for
Quantity
  • Availability
  • Add to Cart