SY3H3D67C1A2 Synthonix Corporation

2,5-Dichloro-2,5-dimethylhexane

  • Cas Number : 6223-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47144.html

Safety Information

MSDS https://synthonix.com/47144.html
Availability details for
Quantity
  • Availability
  • Add to Cart