SY3H3D67C1A4 Synthonix Corporation

2,3-Dimethylcyclohexanol

  • Cas Number : 1502-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47156.html

Safety Information

MSDS https://synthonix.com/47156.html
Availability details for
Quantity
  • Availability
  • Add to Cart