SY3H3D67C1AA Synthonix Corporation

(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine

  • MDL number MFCD09952017
  • Cas Number : 894493-95-9
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1S,2S)-N~1~,N~1~-dimethyl-1,2-cyclohexanediamine
Inchi Code 1S/C8H18N2/c1-10(2)8-6-4-3-5-7(8)9/h7-8H,3-6,9H2,1-2H3/t7-,8-/m0/s1
Inchi Key FRDZGSBXKJXGNR-YUMQZZPRSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Pale-yellow to Brown Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/47196.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47196.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart