SY3H3D67C1B4 Synthonix Corporation

5,6-Dichloro-1H-indazole-3-carbonitrile

  • MDL number MFCD07371566
  • Cas Number : 885278-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47263.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47263.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart