SY3H3D67C1CA Synthonix Corporation

2,8-Dichloroquinazoline

  • MDL number MFCD13193682
  • Cas Number : 67092-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47334.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47334.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart