SY3H3D67C1D7 Synthonix Corporation

3,4-Difluorophenylglyoxal hydrate

  • MDL number MFCD03453081
  • Cas Number : 79784-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white to red powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47475.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47475.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart