SY3H3D67C1D8 Synthonix Corporation

3,5-Difluoro-4-formylbenzoic acid

  • Cas Number : 736990-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/47493.html

Safety Information

MSDS https://synthonix.com/47493.html
Availability details for
Quantity
  • Availability
  • Add to Cart