SY3H3D67C1DF Synthonix Corporation

4,6-Dichloro-2-methylpyrido[3,4-d]pyrimidine

  • MDL number MFCD11044721
  • Cas Number : 1029720-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47635.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47635.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart