SY3H3D67C1EA Synthonix Corporation

2-(2,3-Dichlorophenyl)acetonitrile

  • MDL number MFCD01861392
  • Cas Number : 3218-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47689.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47689.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart