SY3H3D67C1ED Synthonix Corporation

2,3-Difluoro-4-methylbenzonitrile

  • MDL number MFCD06660243
  • Cas Number : 508203-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47701.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47701.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart