SY3H3D67C1F2 Synthonix Corporation

4,5-Difluoro-2-methylbenzonitrile

  • MDL number MFCD09996935
  • Cas Number : 1003708-82-4
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/47706.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/47706.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart