SY3H3D67C204 Synthonix Corporation

1,4-Dibromo-2,5-bis(bromomethyl)benzene

  • Cas Number : 35335-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/47846.html

Safety Information

MSDS https://synthonix.com/47846.html
Availability details for
Quantity
  • Availability
  • Add to Cart