SY3H3D67C213 Synthonix Corporation

4,5-Dichloro-2-methylbenzoic acid

  • MDL number MFCD22566295
  • Cas Number : 5252-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48163.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48163.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart