SY3H3D67C219 Synthonix Corporation

2,3-Difluoro-4-methylbenzoic acid

  • MDL number MFCD01631643
  • Cas Number : 261763-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Off white to Light-gray Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48191.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48191.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart