SY3H3D67C28C Synthonix Corporation

2,4-Difluoro-3-methylbenzaldehyde

  • MDL number MFCD07777152
  • Cas Number : 847502-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to Pale-yellow to Yellow-brown Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48288.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48288.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart