SY3H3D67C28E Synthonix Corporation

3,5-Diiodo-4-methylbenzaldehyde

  • Cas Number : 861118-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/48293.html

Safety Information

MSDS https://synthonix.com/48293.html
Availability details for
Quantity
  • Availability
  • Add to Cart