SY3H3D67C296 Synthonix Corporation

2,2-Dihydroxy-1-(4-nitrophenyl)ethanone

  • Cas Number : 4996-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48540.html

Safety Information

MSDS https://synthonix.com/48540.html
Availability details for
Quantity
  • Availability
  • Add to Cart