SY3H3D67C29C Synthonix Corporation

1,5-Dibromo-2,4-dimethylbenzene

  • MDL number MFCD00154984
  • Cas Number : 615-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Off white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48755.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48755.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart