SY3H3D67C2A3 Synthonix Corporation

3,4-Dihydroquinazolin-2(1H)-one

  • Cas Number : 66655-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48807.html

Safety Information

MSDS https://synthonix.com/48807.html
Availability details for
Quantity
  • Availability
  • Add to Cart