SY3H3D67C2A8 Synthonix Corporation

5,7-Dimethyl-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD00462463
  • Cas Number : 87253-62-1
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/48871.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/48871.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart