SY3H3D67C2B2 Synthonix Corporation

2,4-Dihydroxy-3-methylbenzaldehyde

  • MDL number MFCD00156883
  • Cas Number : 6248-20-0
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Light-red or Yellow Brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49103.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49103.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart