SY3H3D67C2BE Synthonix Corporation

2,3-Difluoroethoxybenzene

  • MDL number MFCD07368737
  • Cas Number : 121219-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49152.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49152.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart