SY3H3D67C326 Synthonix Corporation

2,6-Dichloro-3,5-dimethoxyaniline

  • MDL number MFCD21648506
  • Cas Number : 872509-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49296.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49296.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart