SY3H3D67C328 Synthonix Corporation

2,3-Dimethyl-4-nitrophenol

  • MDL number MFCD01861284
  • Cas Number : 19499-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Slightly pale yellow - Yellow green
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49309.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49309.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart